Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
30µG/G AG, AL, B, BA, CA, CD, CR, CU, FE, MG, MN, MO, NA, NI, P, PB, SI, SN, TI, V, ZN IN HYDROCARBON OIL 1 * 50 g
Catalog # PROOVHG-V21-30-50G
Supplier:
LGC Standards PROMOCHEM
Expand image
30µG/G AG, AL, B, BA, CA, CD, CR, CU, FE, MG, MN, MO, NA, NI, P, PB, SI, SN, TI, V, ZN IN HYDROCARBON OIL 1 * 50 g
Catalog # PROOVHG-V21-30-50G
Supplier:
LGC Standards PROMOCHEM