Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
**USE VHG-9150-01-307-3343** D12-005 STANDARD: AG, AL, CR, CU, FE, MG, NA, NI, PB, SI, SN, TI @ 5 µG/G 1 * 200 g
Catalog # PROOVHG-D12-5-200G
Supplier:
LGC Standards PROMOCHEM
Expand image
**USE VHG-9150-01-307-3343** D12-005 STANDARD: AG, AL, CR, CU, FE, MG, NA, NI, PB, SI, SN, TI @ 5 µG/G 1 * 200 g
Catalog # PROOVHG-D12-5-200G
Supplier:
LGC Standards PROMOCHEM