Nitromethane 96% ACS
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:75-52-5
- Formula:CH₃NO₂
- MDL Number:MFCD00007400
- Molecular Weight:61.04 g/mol
- Merck Index:13,06644
- Boiling Pt:101.2 °C (1013 hPa)
- Melting Pt:–29 °C
- Density:1.12 g/cm³
- Flash Pt:35 °C
- Storage Temperature:Ambient
- EINECS:200-876-6
- UN:1261
- ADR:3,II
Documents
ACRO42401
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3805 to 1.3825 (20°C, 589 nm)
- Water:≤0.05 % (Coulometric)
- GC:≥95 %
- Color scale:≤10 APHA
Specifications
- Pack type:Glass bottle
- Pk:1 L



