4-Hydroxybenzaldehyde 99%
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
4-Hydroxybenzaldehyde 99%
Catalog # ACRO162770025
Supplier: Thermo Fisher Scientific
CAS Number: 123-08-0
Jump to:
Product Details & Documents
- CAS Number:123-08-0
- Formula:C₇H₆O₂
- MDL Number:MFCD00006939
- Molecular Weight:122.12 g/mol
- Merck Index:13,04836
- Boiling Pt:310 °C (1013 hPa)
- Melting Pt:114…118 °C
- Density:1,13 g/cm³ (20 °C)
- Flash Pt:174 °C
- Storage Temperature:Ambient
- EINECS:204-599-1
Documents
ACRO16277
SDS
Specifications
Specification Test Results
- Appearance:Yellow to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:114 to 118 °C
- GC:≥98.5 %
- Stabilizer:2 to 5 ppm BHT
- Residue after evaporation:≤0.001 %
- Acidity (CH3COOH):≤0.002
- Peroxides:≤0.005 % (as H2O2)
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg
- Cat. No.:ACRO162770025
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



