Nitromethane 99+% for analysis
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Nitromethane 99+% for analysis
Catalog # ACRO213370025
Supplier: Thermo Fisher Scientific
CAS Number: 75-52-5
Jump to:
Product Details & Documents
- CAS Number:75-52-5
- Formula:CH₃NO₂
- MDL Number:MFCD00007400
- Molecular Weight:61.04 g/mol
- Merck Index:13,06644
- Boiling Pt:100…102 °C (760 mmHg)
- Melting Pt:–29 °C
- Density:1.12 g/cm³
- Flash Pt:35 °C
- Storage Temperature:Ambient
- EINECS:200-876-6
- UN:1261
- ADR:3,II
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3805 to 1.3825 (20°C, 589 nm)
- Water:≤0.5 % (Coulometric)
- GC:≥99.0 %
- Residue after evaporation:≤0.005 %
- Cadmium (Cd):≤0.1 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.5 ppm
- Lead (Pb):≤0.1 ppm
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



