Pyrazine 99+%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:290-37-9
- Formula:C₄H₄N₂
- MDL Number:MFCD00006122
- Molecular Weight:80.09 g/mol
- Merck Index:13,08047
- Boiling Pt:115…116 °C (760 mmHg)
- Melting Pt:52…56 °C
- Density:1,0311 g/cm³ (60.9 °C)
- Flash Pt:55 °C
- Storage Temperature:Ambient
- EINECS:206-027-6
- UN:1325
- ADR:4.1,III
Documents
ACRO13170
SDS
Specifications
Specification Test Results
- Appearance:White Crystals
- Infrared spectrum:Conforms
- Melting point:52 to 56 °C
- GC:≥99.0 %
Specifications
- Pack type:Glass bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



