(±)-2-Octanol 97%
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol
- CAS Number:123-96-6
- Formula:C₈H₁₈O
- MDL Number:MFCD00004591
- Molecular Weight:130.23 g/mol
- Merck Index:13,06785
- Boiling Pt:174…181 °C (760 mmHg)
- Melting Pt:–38 °C
- Density:0.81 g/cm³
- Flash Pt:72 °C
- Storage Temperature:Ambient
- EINECS:204-667-0
- ADR:6.1,III
Documents
ACRO12941
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4250 to 1.4270 (20°C, 589 nm)
- GC:≥96.0 %
- Titration with NaOH:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



