Propionic acid 99%, pure
Plastic drum, 10 L
Supplier: Thermo Fisher Scientific
Propionic acid 99%, pure
Catalog # ACRO149300100
Supplier: Thermo Fisher Scientific
CAS Number: 79-09-4
Jump to:
Product Details & Documents
Synonyms: Propanoic acid
- CAS Number:79-09-4
- Formula:C₃H₆O₂
- MDL Number:MFCD00002756
- Molecular Weight:74.08 g/mol
- Merck Index:13,07917
- Boiling Pt:141 °C (760 mmHg)
- Melting Pt:–22 °C
- Density:0.99 g/cm³
- Flash Pt:51 °C
- Storage Temperature:Ambient
- EINECS:201-176-3
- UN:3463
- ADR:8,II
- REACH:01-2119486971-24
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3850 to 1.3870 (20°C, 589 nm)
- Water:≤0.2 %
- GC:≥98.5 %
- Color scale:≤10 APHA
Specifications
- Pack type:Plastic drum
- Pk:10 L



