Order Entry
Finland
Orders LinkContactUsLinkComponent
Nitromethane 99%, pure

Nitromethane 99%, pure

Metal drum, 10 L
Catalog # ACRO148510100
Flammable
Irritant
Health
Nitromethane 99%, pure
Catalog # ACRO148510100
CAS Number:  75-52-5
Nitromethane 99%, pure
Nitromethane 99%, pure

Metal drum, 10 L

CAS Number:  75-52-5
Price
1 140,00 €
Log in for availability
1 * 10 L
Product Details & Documents
  • CAS Number:75-52-5
  • Formula:CH₃NO₂
  • MDL Number:MFCD00007400
  • Molecular Weight:61.04 g/mol
  • Merck Index:13,06644
  • Boiling Pt:100…102 °C (760 mmHg)
  • Melting Pt:–29 °C
  • Density:1.12 g/cm³
  • Flash Pt:35 °C
  • Storage Temperature:Ambient
  • EINECS:200-876-6
  • UN:1261
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3810 to 1.3830 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤20 APHA

Specifications

  • Pack type:
    Metal drum
  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.