Fumaric acid 99+%
Plastic drum, 10 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: trans-1,2-Ethylentricarboxylic acid
- CAS Number:110-17-8
- Formula:C₄H₄O₄
- MDL Number:MFCD00002700
- Molecular Weight:116.07 g/mol
- Merck Index:13,04308
- Boiling Pt:356 °C (1013 hPa)
- Melting Pt:295…300 °C (subl.)
- Density:1.62 g/cm³
- Flash Pt:230 °C
- Storage Temperature:Ambient
- EINECS:203-743-0
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:295 to 300 °C
- Water:≤0.25 % (K.F.)
- HPLC:≥99.0 %
- Titration with NaOH:99.0 to 101.0 % (On anhydrous substance)
Specifications
- Pack type:Plastic drum
- Pk:10 kg



