Order Entry
Finland
ContactUsLinkComponent
MicroKros 41,5cm MPES 3kD 1,0mm MLL X FLL sterile 1 * 1 items
Catalog # SPECC04-E003-10-S
MicroKros 41,5cm MPES 3kD 1,0mm MLL X FLL sterile 1 * 1 items
Catalog # SPECC04-E003-10-S
Frequently Bought Together