Accessories for multi-mode Varioskan™ reader
TRF emission filter, 520 nm HBW10 D25 mm
Catalog # LABLF52010
Supplier: THERMO LABSYSTEMS LIFE SCIENCE
Accessories for multi-mode Varioskan™ reader
Catalog # LABLF52010
Supplier: THERMO LABSYSTEMS LIFE SCIENCE
Jump to:
Product Details & Documents
Specifications
- Description:TRF emission filter, 520 nm HBW10 D25 mm
- Seq:21