Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Ultra II Aqueous C18 3um 50x2.1mm 1 * 1 items
Catalog # REST9608352
Supplier:
Restek
Expand image
Ultra II Aqueous C18 3um 50x2.1mm 1 * 1 items
Catalog # REST9608352
Supplier:
Restek