Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Pinnacle DB C1 1.9um 50x2.1mm 1 * 1 items
Catalog # REST9411252
Supplier:
Restek
Expand image
Pinnacle DB C1 1.9um 50x2.1mm 1 * 1 items
Catalog # REST9411252
Supplier:
Restek