Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Grounding plug EU 1 megohm, for connection to building services, 3x 10 mm press button 1 * 1 items
Catalog # SCTV108186
Supplier:
S.C.A.T.
Expand image
Grounding plug EU 1 megohm, for connection to building services, 3x 10 mm press button 1 * 1 items
Catalog # SCTV108186
Supplier:
S.C.A.T.
Related Information
Documentation
Application Notes
GHS Poster
Important Lab and HPLC Safety Guidelines
Documentation