Media in Bottles
TSB Tryptic soy broth, —, 750 ml, Without cap, 1000 ml, 6
Catalog # HEIP1463420006
Supplier: MERCK LIFE SCIENCE
Jump to:
Product Details & Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:6
- Standards:—
- Description:TSB Tryptic soy broth
- Size:1000 ml
- Closure type:Without cap
- Filling:750 ml



