Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Settle plates, single-bagged

Sabouraud, LI, EP, 30 ml, 120
Catalog # HEIP1460030120
Supplier:   MERCK LIFE SCIENCE
Settle plates, single-bagged
Catalog # HEIP1460030120
Supplier:  MERCK LIFE SCIENCE
CAS Number:  
Settle plates, single-bagged
Settle plates, single-bagged

Sabouraud, LI, EP, 30 ml, 120

CAS Number:  
Price
361,00 €
Log in for availability
1 * 120 Items
Product Details & Documents

A wide range of ready-to-use culture media for the environmental monitoring. 90 mm LI (Long Incubation) plates for the determination of the total cell count and for the test for specified microorganisms

  • Designed for the environmental monitoring in non critical areas
Specifications

Specifications

  • Pk:
    120
  • Standards:
    EP
  • Description:
    Sabouraud, LI
  • Filling:
    30 ml

Specifications