Order Entry
Finland
Orders LinkContactUsLinkComponent
Settle plates, single-bagged

Settle plates, single-bagged

TSA with LTHth, LI, -, 30 ml, 120
Catalog # HEIP1460020120
Supplier:   MERCK LIFE SCIENCE
Settle plates, single-bagged
Catalog # HEIP1460020120
Supplier:  MERCK LIFE SCIENCE
CAS Number:  
Settle plates, single-bagged
Settle plates, single-bagged

TSA with LTHth, LI, -, 30 ml, 120

CAS Number:  
Price
360,00 €
Log in for availability
1 * 120 Items
Product Details & Documents

A wide range of ready-to-use culture media for the environmental monitoring. 90 mm LI (Long Incubation) plates for the determination of the total cell count and for the test for specified microorganisms

  • Designed for the environmental monitoring in non critical areas

Documents

Specifications

Specifications

  • Pk:
    120
  • Standards:
    -
  • Description:
    TSA with LTHth, LI
  • Filling:
    30 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.