Order Entry
Finland
Orders LinkContactUsLinkComponent
Data Logger, Einstein LabMate
Data Logger, Einstein LabMate
Data logger for science and education, einstein™ LabMate
Catalog # FOURENLM
Supplier:   FOURIER SYSTEMS
Data Logger, Einstein LabMate
Catalog # FOURENLM
Supplier:  FOURIER SYSTEMS
Data Logger, Einstein LabMate
Data logger for science and education, einstein™ LabMate
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
The LabMate + is a science, technology engineering and mathematics education tool, ideal for primary and secondary schools already equipped with tablets or computers.
  • Small and durable enough to put in a terrarium, or take in the pocket for outdoor science exploration
  • Range of six built-in sensors plus the connectivity with four inputs for a range of optional external sensors (eight sensors possible with splitters)
  • Runs experiment over 24 hours without charging
Pairs to any device via Bluetooth or a USB cable. Built-in sensors include: Heart Rate Sensor, Ambient Humidity Sensor, Light Sensor, UV Sensor, Pressure Sensor, Ambient Temperature.
Ordering information: Pressure kit must be ordered separately.
Specifications
  • Description:
    Data logger for science and education, einstein™ LabMate
  • Cat. No.:
    FOURENLM
  • Interface:
    USB
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Sensor adapter
    Sensor adapter
    FOUREN103
    Log in for availability
    1 * 1 Items
Product Information
  • Catalog #
    Availability
    Price
  • Blood pressure sensor for einstein™ LabMate
    Blood pressure sensor for einstein™ LabMate
    FOURENBLD098
    Log in for availability
    1 * 1 Items
Product Information
  • Catalog #
    Availability
    Price
  • Sensor cable
    Sensor cable
    FOURENCBL022
    Log in for availability
    1 * 1 Items