Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Viscosity Standards(cSt) reads 1.3@20C, 1.2@25C,
[email protected]
, 0.97@40C, 0.87@50C 1 * 500 mL
Catalog # REGEREVIS-N1.0
Supplier:
REAGECON
Expand image
Viscosity Standards(cSt) reads 1.3@20C, 1.2@25C,
[email protected]
, 0.97@40C, 0.87@50C 1 * 500 mL
Catalog # REGEREVIS-N1.0
Supplier:
REAGECON