Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Viscosity Standards(cSt) reads 77000@20C, 47000@25C, 13000@40C, 6100@50C 1 * 500 mL
Catalog # REGEREVIS-N15000
Supplier:
REAGECON
Expand image
Viscosity Standards(cSt) reads 77000@20C, 47000@25C, 13000@40C, 6100@50C 1 * 500 mL
Catalog # REGEREVIS-N15000
Supplier:
REAGECON