0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
DENSITY STANDARD 1.9108G/ML @ 25C 1 * 100 mL
REGEDEN25230PY
:
REAGECON
DENSITY STANDARD 1.9108G/ML @ 25C 1 * 100 mL
REGEDEN25230PY
:
REAGECON