Media in Bottles
Nacl Peptone buffered with LTH, EP, 90 ml, Without cap, 275 ml, 6
Supplier: MERCK LIFE SCIENCE
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:6
- Standards:EP
- Description:Nacl Peptone buffered with LTH
- Size:275 ml
- Closure type:Without cap
- Filling:90 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



