Order Entry
Finland
ContactUsLinkComponent
Heating Mantles WHM, for 20 L reaction vessels with bottom outlet, cylindrical, A-Ø 290 mm, Height 420 mm, without power controller, temperature range 450°C,2 kW, steel casing, power supply 230 V 1 * 1 items
Catalog # WITGDH.WHM122381
Supplier:  witeg Labortechnik
Heating Mantles WHM, for 20 L reaction vessels with bottom outlet, cylindrical, A-Ø 290 mm, Height 420 mm, without power controller, temperature range 450°C,2 kW, steel casing, power supply 230 V 1 * 1 items
Catalog # WITGDH.WHM122381
Supplier:  witeg Labortechnik