Order Entry
Finland
ContactUsLinkComponent
HEATING MANTLES WHM, FOR 2000 ML REACTION VESSELS W. BOTTOM OUTLET, CYLINDRICAL, A-Ø 130 MM, HEIGHT 190 MM, WITHOUT POWER CONTROLLER, TEMPERATURE RANGE 450°C, 450 W, ALUMINIUM CASING, POWER SUPPLY 230 V SERIAL NO. 1 * 1 items
Catalog # WITGDH.WHM12233
Supplier:  witeg Labortechnik
HEATING MANTLES WHM, FOR 2000 ML REACTION VESSELS W. BOTTOM OUTLET, CYLINDRICAL, A-Ø 130 MM, HEIGHT 190 MM, WITHOUT POWER CONTROLLER, TEMPERATURE RANGE 450°C, 450 W, ALUMINIUM CASING, POWER SUPPLY 230 V SERIAL NO. 1 * 1 items
Catalog # WITGDH.WHM12233
Supplier:  witeg Labortechnik