Diffraction gratings
Diaphragm with 4 multiple slits, Slit spacing: 0,25 mm, Slit width: 0, 15 mm
Catalog # BINHU14102
Marketsource
Supplier: 3B Scientific
Jump to:
Product Details & Documents
Photographic diffraction objects in slide frames.
- Dimensions: 50×50 mm
Specifications
- Description:Diaphragm with 4 multiple slits, Slit spacing: 0,25 mm, Slit width: 0, 15 mm