Order Entry
Finland
Orders LinkContactUsLinkComponent
Diffraction gratings
Diffraction gratings
Diaphragm with 3 double slits, Slit spacing: 0,30 mm, Slit width: 0,10 mm; 0,15 mm; 0,20 mm
Catalog # BINHU14100
Marketsource
Supplier:   3B Scientific
Diffraction gratings
Catalog # BINHU14100
Supplier:  3B Scientific
Marketsource
Diffraction gratings
Diffraction gratings
Diaphragm with 3 double slits, Slit spacing: 0,30 mm, Slit width: 0,10 mm; 0,15 mm; 0,20 mm
Price
35,90 €
Log in for availability
1 * 1 Items
Product Details & Documents
Photographic diffraction objects in slide frames.
  • Dimensions: 50×50 mm
Specifications
  • Description:
    Diaphragm with 3 double slits, Slit spacing: 0,30 mm, Slit width: 0,10 mm; 0,15 mm; 0,20 mm