Media in Bottles
TSB Tryptic soy broth, EP, USP, 100 ml, Cap with septum, 125 ml, 10
Supplier: MERCK LIFE SCIENCE
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:10
- Standards:EP, USP
- Description:TSB Tryptic soy broth
- Size:125 ml
- Closure type:Cap with septum
- Filling:100 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



