Order Entry
Finland
Orders LinkContactUsLinkComponent
Settle plates, single-bagged

Settle plates, single-bagged

TSA, LI, EP, USP, 30 ml, 20
Catalog # HEIP1460040020
Supplier:   MERCK LIFE SCIENCE
Settle plates, single-bagged
Catalog # HEIP1460040020
Supplier:  MERCK LIFE SCIENCE
CAS Number:  
Settle plates, single-bagged
Settle plates, single-bagged

TSA, LI, EP, USP, 30 ml, 20

CAS Number:  
Price
78,00 €
Log in for availability
1 * 20 Items
Product Details & Documents

A wide range of ready-to-use culture media for the environmental monitoring. 90 mm LI (Long Incubation) plates for the determination of the total cell count and for the test for specified microorganisms

  • Designed for the environmental monitoring in non critical areas

Documents

Specifications

Specifications

  • Pk:
    20
  • Standards:
    EP, USP
  • Description:
    TSA, LI
  • Filling:
    30 ml

Specifications