SPE columns, Supra-Poly® HLB
HLB, 30 µm, 3 ml, 30 mg
PERCN9306651
- Contains macro-porous polymers with ultra-pure, pharmaceutical grade spherical particles
- Shorter analysis times, greater load capacity and reduced solvent usage
- Ideal for high throughput assays
- Particle Size:30 µm
- Capacity:3 ml
- Seq:05
- Bed weight:30 mg
- Phase:HLB
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



