Order Entry
Finland
ContactUsLinkComponent
HGF Human Recombinant produced in CHO is heterodimer, non-glycosylated, polypeptide chain consisting an alpha chain of 463 amino aci ds and b-chain of 234 having total molecular mass of 75kDa.The HGF is purified by proprietary chromatographi 1 * 2 µG
Catalog # BIRBORB80379-2
Supplier:  Biorbyt
HGF Human Recombinant produced in CHO is heterodimer, non-glycosylated, polypeptide chain consisting an alpha chain of 463 amino aci ds and b-chain of 234 having total molecular mass of 75kDa.The HGF is purified by proprietary chromatographi 1 * 2 µG
Catalog # BIRBORB80379-2
Supplier:  Biorbyt
Related Information