Order Entry
Finland
ContactUsLinkComponent
Placental Growth Hormone Ovine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 191 amino a cids and having a molecular mass of 21918 Dalton. The GH Ovine Recombinant is purified by proprietary chroma 1 * 1 mg
Catalog # BIRBORB80357-1
Supplier:  Biorbyt
Placental Growth Hormone Ovine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 191 amino a cids and having a molecular mass of 21918 Dalton. The GH Ovine Recombinant is purified by proprietary chroma 1 * 1 mg
Catalog # BIRBORB80357-1
Supplier:  Biorbyt
Related Information