Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
C18G x 3µm particle size x 300Å pore size, 150mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C184-03N02S
Supplier:
Trajan Scientific and Medical
Expand image
C18G x 3µm particle size x 300Å pore size, 150mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C184-03N02S
Supplier:
Trajan Scientific and Medical