Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Silica x 5µm particle size x 120Å pore size, 100mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
SGEA2SIL-05M46S
:
Trajan Scientific and Medical
Silica x 5µm particle size x 120Å pore size, 100mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
SGEA2SIL-05M46S
:
Trajan Scientific and Medical