Insole, Action Dry
Insoles, Action Dry, 39, Black
Catalog # UPOWCS00015/39
Supplier: U-Power
Jump to:
Product Details & Documents
Made from EVA/textile.
- Light non-slip
- Keeps feet dry
- Anti-fungal treated
Specifications
- Shoe size:39
- Colour:Black
- Description:Insoles, Action Dry