Tablet, einstein tablet
einstein™ Tablet +
Catalog # FOURENTABSEN
Supplier: FOURIER SYSTEMS
Jump to:
Product Details & Documents
The einstein ™ Tablet + is a full Android tablet with a built-in science lab designed for primary and secondary science, technology engineering and mathematics education (K-12 STEM).
- Contains 8 built-in sensors, commonly used in most science curricula, and connects to up to 8 external sensors
- Designed to be used both in the science lab and also in a broader educational environment
- Free apps and software included
Built-in sensors include:
Temperature
Humidity
Microphone
Light
UV
Heart rate - jack for external sensor
GPS
Accelerometer
Specifications
- Description:einstein™ Tablet +