Order Entry
Finland
ContactUsLinkComponent
Biotix 250ul clear pipet tips for Beckman 96-well workstations. Racked, 10 racks of 96 Tips/Pack, 5 Packs/Case. AP96 Tips for Beck man Biomek FX/NX, MultiMek 96, and Biomek 3000 workstations. Presterilized, RNase-, DNase-, and Endotoxin (pyrogen)- 1 * 4.800 items
Catalog # 732-2453
Supplier:  BIOTIX
Biotix 250ul clear pipet tips for Beckman 96-well workstations. Racked, 10 racks of 96 Tips/Pack, 5 Packs/Case. AP96 Tips for Beck man Biomek FX/NX, MultiMek 96, and Biomek 3000 workstations. Presterilized, RNase-, DNase-, and Endotoxin (pyrogen)- 1 * 4.800 items
Catalog # 732-2453
Supplier:  BIOTIX
Frequently Bought Together