Order Entry
Finland
ContactUsLinkComponent
pH combination electrode with Memosens screw plug head (ATEX II 1/G) low maintenance, glass shaft, KPG annular gap junction, Referid -electrolyte, Silamid-reference system, dome membrane, H-glass, length 360 mm, 12 mm Ø, 0...+100 °C, 2...13 pH 1 * 1 items
Catalog # SCOI285111030
Supplier:  SI Analytics
pH combination electrode with Memosens screw plug head (ATEX II 1/G) low maintenance, glass shaft, KPG annular gap junction, Referid -electrolyte, Silamid-reference system, dome membrane, H-glass, length 360 mm, 12 mm Ø, 0...+100 °C, 2...13 pH 1 * 1 items
Catalog # SCOI285111030
Supplier:  SI Analytics