Rechargable Batteries
Rechargable NiMH Power ACCU™, Micro AAA/HR03, 1,2 V, 1000 mAh
Catalog # 139-0603
Supplier: VARTA GERAETEBATTERIE
Jump to:
Product Details & Documents
High capacity with long operating times per charge. Do not require a special charger.
- Can be recharged up to 1000 times
Specifications
- Type:Micro AAA/HR03
- Voltage:1,2 V
- Capacity:1000 mAh
- Description:Rechargable NiMH Power ACCU™
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



