High quality prismatic optical bench
Simple rider for triangular bench, column height 80 mm, for rods up to 10 mm diameter
DIDAPOD060080
: DIDALAB
Triangular, cast aluminium optical bench
- Lightweight for easy transport and manoeuvrability
- Graduated measure to accurately determine distance between components for reproducibility of results
- Compatible with most triangular riders
- Adjustable feet for stability
- Description:Simple rider for triangular bench, column height 80 mm, for rods up to 10 mm diameter