High quality prismatic optical bench
Rider for triangular bench for vertical (25 mm) and horizontal (±50 mm) motion
Catalog # DIDAPOD060110
Supplier: DIDALAB
Jump to:
Product Details & Documents
Triangular, cast aluminium optical bench
- Lightweight for easy transport and manoeuvrability
- Graduated measure to accurately determine distance between components for reproducibility of results
- Compatible with most triangular riders
- Adjustable feet for stability
Specifications
- Description:Rider for triangular bench for vertical (25 mm) and horizontal (±50 mm) motion