Order Entry
Finland
ContactUsLinkComponent
Human AMID partial ORF ( AAH06121, 1 A.A. - 339 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00084883-Q022
Supplier:  Abnova
Human AMID partial ORF ( AAH06121, 1 A.A. - 339 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00084883-Q022
Supplier:  Abnova