Order Entry
Finland
ContactUsLinkComponent
Neptune 1000 µl pipette tips. environmentally sustainable pack (esp) reload system, pre-sterile, 10 cards of 96 tips/pack, 5 packs/c ase. universal fit ideal for both single and multichannel pipettors, graduation marks. 1 * 960 items
Catalog # BITX2167.96.S
Supplier:  BIOTIX
Neptune 1000 µl pipette tips. environmentally sustainable pack (esp) reload system, pre-sterile, 10 cards of 96 tips/pack, 5 packs/c ase. universal fit ideal for both single and multichannel pipettors, graduation marks. 1 * 960 items
Catalog # BITX2167.96.S
Supplier:  BIOTIX
Frequently Bought Together