Order Entry
Finland
ContactUsLinkComponent
Neptune 10 µl pipettetips. pre-sterile, racked, 10 racks of 96 tips/pack, 5 packs/case. universal fit ideal for both single and mul tichannel pipettors. 1 * 960 items
Catalog # BITX2042.NS
Supplier:  BIOTIX
Neptune 10 µl pipettetips. pre-sterile, racked, 10 racks of 96 tips/pack, 5 packs/case. universal fit ideal for both single and mul tichannel pipettors. 1 * 960 items
Catalog # BITX2042.NS
Supplier:  BIOTIX
Frequently Bought Together