Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Kirjaudu sisään
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Lysing matrix b 1 * 100 items
Catalog # ICNA116911100
Supplier:
MP Biomedicals
Expand image
Lysing matrix b 1 * 100 items
Catalog # ICNA116911100
Supplier:
MP Biomedicals