Order Entry
Finland
ContactUsLinkComponent
Double position ultra clean base plate for gc. a base plate allows cartridges to be exchanged without introducing oxygen. its featur es and benefits include - ultra-high capacity high-purity output ensures 99.9999% pure gas quick connect fittings f 1 * 1 items
Catalog # PERCN9306805
Supplier:  PerkinElmer
Double position ultra clean base plate for gc. a base plate allows cartridges to be exchanged without introducing oxygen. its featur es and benefits include - ultra-high capacity high-purity output ensures 99.9999% pure gas quick connect fittings f 1 * 1 items
Catalog # PERCN9306805
Supplier:  PerkinElmer