ISE standards
Sodium standard solution, 1000 mg/l (500 ml), 1000 mg/l as Na, 1
Supplier: Hach
Jump to:
Product Details & Documents
ISE standards are used to calibrate ion selective electrode measurement systems by coresponding the electrode output with the concentration of the analyte. Pairs of standards with concentrations that differ by a factor of ten are used for analysis.
Documents
Specifications
Specifications
- Concentration:1000 mg/l as Na
- Pk:1
- Description:Sodium standard solution, 1000 mg/l (500 ml)
Specifications
Accessories
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Sodium standard solution, 100 mg/lSodium standard solution, 100 mg/lHACH2318149100 mg/l as Na500 mlLog in for availability1 * 500 mL - 69,00 €
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Nitrogen-nitrate standard solution, 10 mg/lNitrogen-nitrate standard solution, 10 mg/lHACH3074910 mg/l as NO₃-N500 mLLog in for availability1 * 500 mL - 46,70 €
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Chloride standard solution, 100 mg/lChloride standard solution, 100 mg/lHACH2370853100 mg/l as Cl1 LLog in for availability1 * 1 L - 72,40 €
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Nitrogen-nitrate standard solution, 1000 mg/lNitrogen-nitrate standard solution, 1000 mg/lHACH12792491000 mg/l as NO₃-N500 mLLog in for availability1 * 500 mL - 53,30 €
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Calcium standard solution, 10 mg/lCalcium standard solution, 10 mg/lHACH230544210 mg/l as Ca100 mLLog in for availability1 * 100 mL - 46,60 €
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Nitrogen-ammonia standard solution, 1000 mg/lNitrogen-ammonia standard solution, 1000 mg/lHACH23541531000 mg/l as NH₃-N1 LLog in for availability1 * 1 L - 47,60 €
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Potassium standard solution, 100 mg/lPotassium standard solution, 100 mg/lHACH2351749100 mg/l as K500 mLLog in for availability1 * 500 mL - 92,80 €
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Fluoride standard solution, 100 mg/lFluoride standard solution, 100 mg/lHACH23249100 mg/l as F500 mLLog in for availability1 * 500 mL - 68,10 €
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Nitrogen-ammonia standard solution, 100 mg/lNitrogen-ammonia standard solution, 100 mg/lHACH2406549100 mg/l as NH₃-N500 mLLog in for availability1 * 500 mL - 46,70 €
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Sodium standard solution, 1000 mg/l (500 ml)Sodium standard solution, 1000 mg/l (500 ml)HACH14749491000 mg/l as Na1Log in for availability1 * 1 Items - 70,80 €