TAPS, 0,2 M buffer soln., pH 8,0
TAPS buffer solution, 0,2 M, 8,0, 100 mL
Supplier: Thermo Fisher Scientific
Siirry kohtaan:
Product Details & Documents
CAS No.: 91000-53-2
Physical Form: Liquid
MDL No.: MFCD00069903
Documents
Tekniset tiedot
Tekniset tiedot
- Concentration:0,2 M
- Pk:100 mL
- Description:TAPS buffer solution
- pH value:8,0
- Cat. No.:J60322.AE
- Physical form:Liquid
- MDL No.:MFCD00069903
- CAS No.:91000-53-2
Tekniset tiedot
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



