Order Entry
Finland
Orders LinkContactUsLinkComponent
AMPD, 0.2 M Buffer Solutions

AMPD, 0.2 M Buffer Solutions

AMPD buffer solution, 0,2 M, 9,5, 100 mL
Catalog # J60916.AE
AMPD, 0.2 M Buffer Solutions
Catalog # J60916.AE
CAS Number:  
AMPD, 0.2 M Buffer Solutions
AMPD, 0.2 M Buffer Solutions

AMPD buffer solution, 0,2 M, 9,5, 100 mL

CAS Number:  
Price
112,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

2-Amino-2-methyl-1,3-propanediol buffer, colorless and odorless.

CAS No.: 115-69-5
MDL No.: MFCD00004678

Caution: Storage Temperature: 2 to 8 °C

Specifications

Specifications

  • Concentration:
    0,2 M
  • Pk:
    100 mL
  • Description:
    AMPD buffer solution
  • pH value:
    9,5
  • Cat. No.:
    J60916.AE
  • Odour/scent:
    Odourless
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00004678
  • Appearance:
    Colourless
  • CAS No.:
    115-69-5

Specifications