Order Entry
Finland
ContactUsLinkComponent
2 channel recalibratable high precision thermometer TD 241 for multi probes, Range: - 200°C...+1750 °C . Can be recalibrate d at 0°C and scale. Supplied with a type K probe in a case with calibration certificate 1 * 1 items
Catalog # 620-1935
Supplier:  VWR Collection
2 channel recalibratable high precision thermometer TD 241 for multi probes, Range: - 200°C...+1750 °C . Can be recalibrate d at 0°C and scale. Supplied with a type K probe in a case with calibration certificate 1 * 1 items
Catalog # 620-1935
Supplier:  VWR Collection
Related Information